2-Chloro-5-fluoroisonicotinaldehyde ≥95%

Supplier: Matrix Scientific

074764-250MG 074764-1G 074764-5G
102538-106EA 150.77 USD
102538-106 102538-108 102538-110
2-Chloro-5-fluoroisonicotinaldehyde ≥95%
2-Chloro-5-fluoroisonicotinaldehyde

Hazard: Irritant

Formula: C₆H₃ClFNO
MDL Number: MFCD04972411
CAS Number: 884494-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR