3-Methoxy-5-methylbenzoic acid ≥95%

Supplier: Matrix Scientific

074762-1G 074762-5G 074762-25G
102538-096EA 92.31 USD
102538-096 102538-098 102664-014
3-Methoxy-5-methylbenzoic acid ≥95%
3-Methoxy-5-methylbenzoic acid

Hazard: Irritant

Formula: C9H10O3
MW: 166.1700 g/mol
MDL Number: MFCD12405314
CAS Number: 62089-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR