Ethyl-N-Cbz-4-piperidineacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl N-Cbz-4-piperidineacetate

074760-1G 074760-5G 074760-10G
102538-082EA 178.46 USD
102538-082 102538-084 102538-086
Ethyl-N-Cbz-4-piperidineacetate ≥95%
Ethyl-N-Cbz-4-piperidineacetate

Hazard: Irritant

Formula: C₁₇H₂₃NO₄
MW: 305.3700 g/mol
MDL Number: MFCD09953323
CAS Number: 80221-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR