(S)-Isopropyl-2-aminopropanoate hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-Isopropyl 2-aminopropanoate hydrochloride, (S)-2-Aminopropionic acid Isopropyl ester hydrochloride, L-Alanine Isopropyl ester hydrochloride, H-Ala-OiPr·HCl, Isopropyl (S)-2-Aminopropionate hydrochloride

074759-1G 074759-5G
102538-076EA 41.54 USD
102538-076 102538-078
(S)-Isopropyl-2-aminopropanoate hydrochloride ≥95%
(S)-Isopropyl-2-aminopropanoate hydrochloride

Hazard: Irritant/Refrigerate

Formula: C₆H₁₃NO₂·HCl
MW: 167.6 g/mol
MDL Number: MFCD08059709
CAS Number: 62062-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR