Methyl-4-amino-2-chlorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-amino-2-chlorobenzoate

074758-1G 074758-5G 074758-10G
102538-070EA 46.15 USD
102538-070 102538-072 102538-074
Methyl-4-amino-2-chlorobenzoate ≥95%
Methyl 4-amino-2-chlorobenzoate

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD03840484
CAS Number: 46004-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR