2-(6-Bromopyridin-3-yl)pyrimidine ≥95%

Supplier: Matrix Scientific

074757-250MG 074757-1G
102538-066EA 826.15 USD
102538-066 102538-068
2-(6-Bromopyridin-3-yl)pyrimidine ≥95%
2-(6-Bromopyridin-3-yl)pyrimidine

Hazard: Irritant

Formula: C₉H₆BrN₃
MDL Number: MFCD09999083
CAS Number: 942189-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR