8-Bromoquinoline-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

074752-250MG 074752-1G 074752-5G
102538-040EA 355.38 USD
102538-040 102538-042 102538-044
8-Bromoquinoline-3-carboxylic acid ≥95%
8-Bromoquinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₂
MW: 252.0641 g/mol
MDL Number: MFCD18375331
CAS Number: 347146-16-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR