tert-Butyl-4-formylbenzylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(Aminomethyl)benzaldehyde, N-BOC protected, tert-Butyl 4-formylbenzylcarbamate

074750-1G 074750-5G 074750-10G
102538-028EA 201.54 USD
102538-028 102538-030 102538-032
tert-Butyl-4-formylbenzylcarbamate ≥95%
tert-Butyl-4-formylbenzylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₇NO₃
MW: 235.2800 g/mol
MDL Number: MFCD06659090
CAS Number: 156866-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR