Tetrahydro-1H-pyrrolizine-7a(5H)acetic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: Tetrahydro-1H-pyrrolizine-7a(5H)-acetic acid hydrochloride

074748-1G 074748-5G
102538-020EA 640 USD
102538-020 102538-022
Tetrahydro-1H-pyrrolizine-7a(5H)acetic acid hydrochloride ≥95%
Tetrahydro-1H-pyrrolizine-7a(5H)acetic acid hydrochloride

Hazard: Irritant

Formula: C₉H₁₆ClNO₂
MW: 205.68 g/mol
MDL Number: MFCD09032985
CAS Number: 124655-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR