5-Amino-2-chlorobenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3-cyanoaniline

074745-1G 074745-5G 074745-10G
102538-002EA 427.69 USD
102538-002 102538-004 102538-006
5-Amino-2-chlorobenzonitrile ≥95%
5-Amino-2-chlorobenzonitrile

Hazard: Irritant

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
MDL Number: MFCD00800459
CAS Number: 35747-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR