Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-benzyloxy-4-methoxy-2-nitrobenzoate

074744-1G 074744-5G
102537-998EA 710.77 USD
102537-998 102538-000
Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate ≥95%
Methyl-5-benzyloxy-4-methoxy-2-nitrobenzoate

Hazard: Irritant

Formula: C₁₆H₁₅NO₆
MW: 317.2900 g/mol
MDL Number: MFCD17168894
CAS Number: 164161-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR