4-Amino-3,5-dibromopyridine ≥95%

Supplier: Matrix Scientific

074743-1G 074743-5G 074743-10G
102537-992EA 90.77 USD
102537-992 102537-994 102537-996
4-Amino-3,5-dibromopyridine ≥95%
4-Amino-3,5-dibromopyridine

Hazard: Irritant

Formula: C₅H₄Br₂N₂
MDL Number: MFCD00234048
CAS Number: 84539-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR