(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine ≥95%

Supplier: Matrix Scientific

Synonyms: Rasagiline, (1R)-N-prop-2-ynylindan-1-amine

074739-1G 074739-5G 074739-10G
102537-968EA 196.92 USD
102537-968 102537-970 102537-972
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine ≥95%
(R)-N-(2-Propynyl)-2,3-dihydroinden-1-amine

Hazard: Irritant

Formula: C₁₂H₁₃N
MW: 171.2383 g/mol
MDL Number: MFCD00866571
CAS Number: 136236-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR