Benzyl-4-iodobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl 4-iodobenzoate

074737-1G 074737-5G 074737-10G
102537-956EA 533.85 USD
102537-956 102537-958 102537-960
Benzyl-4-iodobenzoate ≥95%
Benzyl-4-iodobenzoate

Hazard: Irritant

Formula: C₁₄H₁₁IO₂
MW: 338.15 g/mol
MDL Number: MFCD18072656
CAS Number: 136618-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR