N-(2,6-Difluoro-3-nitrophenyl)acetamide ≥95%

Supplier: Matrix Scientific

074736-1G 074736-5G
102537-952EA 412.31 USD
102537-952 102537-954
N-(2,6-Difluoro-3-nitrophenyl)acetamide ≥95%
N-(2,6-Difluoro-3-nitrophenyl)acetamide

Hazard: Irritant

Formula: C₈H₆F₂N₂O₃
MW: 216.1400 g/mol
MDL Number: MFCD00174326
CAS Number: 25892-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR