2-(Piperidin-3-yl)acetic acid hydrochloride ≥95%

Supplier: Matrix Scientific

074735-1G 074735-5G 074735-10G
102537-946EA 249.23 USD
102537-946 102537-948 102537-950
2-(Piperidin-3-yl)acetic acid hydrochloride ≥95%
2-(Piperidin-3-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClNO₂
MW: 179.6446 g/mol
MDL Number: MFCD02181158
CAS Number: 71985-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR