8-Chloro-2-methoxy-1,5-naphthyridine ≥95%

Supplier: Matrix Scientific

074731-250MG 074731-1G
102537-924EA 967.69 USD
102537-924 102537-926
8-Chloro-2-methoxy-1,5-naphthyridine ≥95%
8-Chloro-2-methoxy-1,5-naphthyridine

Hazard: Irritant

Formula: C₉H₇ClN₂O
MDL Number: MFCD09027618
CAS Number: 249889-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR