4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine ≥95%

Supplier: Matrix Scientific

074729-250MG 074729-1G
102537-916EA 450.42 USD
102537-916 102537-918
4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine ≥95%
4-Chloro-3-iodo-1H-pyrazolo[4,3-c]pyridine

Hazard: Irritant

Formula: C₆H₃ClIN₃
MW: 279.4700 g/mol
MDL Number: MFCD16877655
CAS Number: 1186647-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR