2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole ≥95%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl 3-(1H-benzo[d]imidazol-2-yl)piperidine-1-carboxylate
074728-250MG
074728-1G
074728-5G
102537-910EA
427.69
USD
102537-910
102537-912
102537-914
2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole ≥95%
2-(N-Boc-piperidin-3-yl)-1H-benzoimidazole
Hazard: Irritant
Formula:
C₁₇H₂₃N₃O₂ MW: 301.38 g/mol |
MDL Number:
MFCD17214278 CAS Number: 1229000-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...