2-Bromo-3-hydroxybenzaldehyde ≥95%
Supplier: Matrix Scientific
074727-5G
074727-25G
102537-906EA
196.92
USD
102537-906
102833-710
2-Bromo-3-hydroxybenzaldehyde ≥95%
2-Bromo-3-hydroxybenzaldehyde
Hazard: Irritant
Formula:
C₇H₅BrO₂ MW: 201.02 g/mol |
MDL Number:
MFCD00092081 CAS Number: 196081-71-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...