3-Fluoro-5-nitrobenzaldehyde ≥95%
Supplier: Matrix Scientific
074726-1G
074726-5G
074726-10G
102537-898EA
427.69
USD
102537-898
102537-900
102537-902
3-Fluoro-5-nitrobenzaldehyde ≥95%
3-Fluoro-5-nitrobenzaldehyde
Hazard: Irritant
Formula:
C₇H₄FNO₃ MW: 169.1100 g/mol |
MDL Number:
MFCD11111000 CAS Number: 108159-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...