5-Bromo-1,2-dimethyl-3-nitrobenzene ≥95%

Supplier: Matrix Scientific

074722-1G 074722-5G 074722-10G
102537-874EA 533.85 USD
102537-874 102537-876 102537-878
5-Bromo-1,2-dimethyl-3-nitrobenzene ≥95%
5-Bromo-1,2-dimethyl-3-nitrobenzene

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
MDL Number: MFCD08690122
CAS Number: 18873-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR