4-Methoxy-2-nitro-1-(trifluoromethyl)benzene ≥95%

Supplier: Matrix Scientific

074721-1G 074721-5G 074721-10G
102537-868EA 533.85 USD
102537-868 102537-870 102537-872
4-Methoxy-2-nitro-1-(trifluoromethyl)benzene ≥95%
4-Methoxy-2-nitro-1-(trifluoromethyl)benzene

Hazard: Irritant

Formula: C₈H₆F₃NO₃
MDL Number: MFCD11848539
CAS Number: 25889-37-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR