2,3-Dimethyl-4-fluoronitrobenzene ≥95%

Supplier: Matrix Scientific

074720-1G 074720-5G 074720-10G
102537-862EA 233.85 USD
102537-862 102537-864 102537-866
2,3-Dimethyl-4-fluoronitrobenzene ≥95%
2,3-Dimethyl-4-fluoronitrobenzene

Hazard: Irritant

Formula: C₈H₈FNO₂
MW: 169.16 g/mol
MDL Number: MFCD05663712
CAS Number: 1736-87-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR