3-(Aminosulfonyl)propanoic acid ≥95%

Supplier: Matrix Scientific

074714-1G 074714-5G
102537-836EA 569.23 USD
102537-836 102537-838
3-(Aminosulfonyl)propanoic acid ≥95%
3-(Aminosulfonyl)propanoic acid

Hazard: Irritant

Formula: C3H7NO4S
CAS Number: 15441-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR