Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Dicyclohexylphosphino-2’,6’-dimethoxybiphenyl, SPhos, Dicyclohexyl(2',6'-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine, 2-Dicyclohexylphosphino-2',6'-dimethoxybiphenyl, 2-Dicyclohexylphosphino-2',6'-dimethoxy-1,1'-biphenyl

074713-1G 074713-5G
102537-830EA 61.54 USD
102537-830 102537-832
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine ≥95%
Dicyclohexyl(2',6'-dimethoxy[1,1'-biphenyl]-2-yl)phosphine

Hazard: Irritant/Air Sensitive

Formula: C₂₆H₃₅O₂P
MW: 410.53 g/mol
Melting Pt: 164…167 °C
MDL Number: MFCD05861611
CAS Number: 657408-07-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR