2-Bromo-4-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

074712-1G 074712-5G
102537-826EA 184.62 USD
102537-826 102537-828
2-Bromo-4-(trifluoromethyl)pyrimidine ≥95%
2-Bromo-4-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₂BrF₃N₂
MW: 226.9820 g/mol
MDL Number: MFCD09802248
CAS Number: 785777-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR