6-Bromo-2-methylthiazolo[5,4-b]pyridine ≥95%

Supplier: Matrix Scientific

074711-250MG 074711-1G 074711-5G
102537-820EA 684.62 USD
102537-820 102537-822 102537-824
6-Bromo-2-methylthiazolo[5,4-b]pyridine ≥95%
6-Bromo-2-methylthiazolo[5,4-b]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂S
MW: 229.1 g/mol
MDL Number: MFCD07375016
CAS Number: 886372-92-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR