(1H-Pyrrol-2-yl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

074710-250MG 074710-1G 074710-5G
102537-814EA 307.69 USD
102537-814 102537-816 102537-818
(1H-Pyrrol-2-yl)methanamine hydrochloride ≥95%
(1H-Pyrrol-2-yl)methanamine hydrochloride

Hazard: Irritant

Formula: C₅H₉ClN₂
MDL Number: MFCD20037390
CAS Number: 1351479-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR