2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile ≥95%

Supplier: Matrix Scientific

074708-1G 074708-5G
102537-806EA 640 USD
102537-806 102537-808
2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile ≥95%
2-(5-Aminopyridin-2-yl)-2-methylpropanenitrile

Hazard: Irritant

Formula: C₉H₁₁N₃
CAS Number: 1226776-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR