Anethole trithione ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Felviten

074707-1G 074707-5G 074707-10G
102537-800EA 355.38 USD
102537-800 102537-802 102537-804
Anethole trithione ≥95%
Anethole trithione

Hazard: Irritant

Formula: C₁₀H₈OS₃
MW: 240.3649 g/mol
MDL Number: MFCD00129751
CAS Number: 532-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR