(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Fluoro-2,4-dinitrophenyl-5-L-alanine amide, N-alpha-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, Propanamide, 2-[(5-fluoro-2,4-dinitrophenyl)amino]-, (2S)-

074705-1G 074705-5G
102537-790EA 353.85 USD
102537-790 102537-792
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide ≥95%
(S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)propanamide

Hazard: Irritant

Formula: C₉H₉FN₄O₅
MW: 272.192 g/mol
MDL Number: MFCD00042049
CAS Number: 95713-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR