2-(Phenylsulfonyl)pyridine ≥95%
Supplier: Matrix Scientific
074704-1G
074704-5G
074704-10G
102537-784EA
60
USD
102537-784
102537-786
102537-788
2-(Phenylsulfonyl)pyridine ≥95%
2-(Phenylsulfonyl)pyridine
Hazard: Irritant
Formula:
C₁₁H₉NO₂S MW: 219.2600 g/mol |
MDL Number:
MFCD09999263 CAS Number: 24244-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...