5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%

Supplier: Matrix Scientific

074701-1G 074701-5G
102537-772EA 353.85 USD
102537-772 102537-774
5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride ≥95%
5-Methyl-1,2,3,4-tetrahydroquinoline hydrochloride

Hazard: Irritant

Formula: C10H14ClN
MW: 183.6800 g/mol
MDL Number: MFCD09026775
CAS Number: 861368-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR