5-Bromo-2-(4-chlorophenyl)pentanoic acid ≥95%

Supplier: Matrix Scientific

074700-250MG 074700-1G
102537-768EA 1223.08 USD
102537-768 102537-770
5-Bromo-2-(4-chlorophenyl)pentanoic acid ≥95%
5-Bromo-2-(4-chlorophenyl)pentanoic acid

Hazard: Irritant

Formula: C₁₁H₁₂BrClO₂
CAS Number: 1017789-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR