Ethyl-2-amino-5-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-amino-5-methoxybenzoate

074696-1G 074696-5G
102537-750EA 640 USD
102537-750 102537-752
Ethyl-2-amino-5-methoxybenzoate ≥95%
Ethyl-2-amino-5-methoxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₃NO₃
MW: 195.22 g/mol
MDL Number: MFCD11111632
CAS Number: 64018-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR