3-tert-Butylaniline ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(tert-Butyl)aniline, m-tert-Butylaniline
074695-1G
074695-5G
102537-746EA
46.15
USD
102537-746
102537-748
3-tert-Butylaniline ≥95%
3-tert-Butylaniline
Hazard: Irritant
Formula:
C₁₀H₁₅N MW: 149.24 g/mol |
MDL Number:
MFCD00125078 CAS Number: 5369-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...