2-(Bromomethyl)-5-(trifluoromethyl)furan ≥95%
Supplier: Matrix Scientific
074694-1G
074694-5G
102537-742EA
420
USD
102537-742
102537-744
2-(Bromomethyl)-5-(trifluoromethyl)furan ≥95%
2-(Bromomethyl)-5-(trifluoromethyl)furan
Hazard: Irritant
Formula:
C₆H₄BrF₃O MW: 228.9946 g/mol |
MDL Number:
MFCD03086219 CAS Number: 17515-77-4 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...