5-Chloro-4-fluoro-2-nitrophenol ≥95%
Supplier: Matrix Scientific
074692-1G
074692-5G
074692-10G
102537-730EA
533.85
USD
102537-730
102537-732
102537-734
5-Chloro-4-fluoro-2-nitrophenol ≥95%
5-Chloro-4-fluoro-2-nitrophenol
Hazard: Irritant
Formula:
C₆H₃ClFNO₃ MW: 191.5400 g/mol |
MDL Number:
MFCD18072578 CAS Number: 65001-79-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...