3-Fluoro-2-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

074691-250MG 074691-1G 074691-5G
102537-724EA 200 USD
102537-724 102537-726 102537-728
3-Fluoro-2-nitrobenzonitrile ≥95%
3-Fluoro-2-nitrobenzonitrile

Hazard: Irritant

Formula: C₇H₃FN₂O₂
MDL Number: MFCD09864664
CAS Number: 1000339-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR