4-Hydroxyindanone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-HYDROXY-2,3-DIHYDRO-1H-INDEN-1-ONE, 4-hydroxyindan-1-one, 4-Hydroxy-1-indanone

074690-1G 074690-5G 074690-10G
102537-718EA 72.31 USD
102537-718 102537-720 102537-722
4-Hydroxyindanone ≥95%
4-Hydroxyindanone

Hazard: Irritant

Formula: C₉H₈O₂
MW: 148.161 g/mol
MDL Number: MFCD00143330
CAS Number: 40731-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR