(3-Fluoro-4-methoxyphenyl)acetonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-4-methoxylphenylacetonitrile
074689-1G
074689-5G
074689-10G
102537-712EA
69.23
USD
102537-712
102537-714
102537-716
(3-Fluoro-4-methoxyphenyl)acetonitrile ≥95%
(3-Fluoro-4-methoxyphenyl)acetonitrile
Hazard: Irritant
Formula:
C₉H₈FNO MW: 165.17 g/mol |
MDL Number:
MFCD00040889 CAS Number: 404-90-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...