1-Methyl-1H-indazol-5-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Hydroxy-1-methyl-1H-indazole

074688-250MG 074688-1G 074688-5G
102537-706EA 230.77 USD
102537-706 102537-708 102537-710
1-Methyl-1H-indazol-5-ol ≥95%
1-Methyl-1H-indazol-5-ol

Hazard: Irritant

Formula: C₈H₈N₂O
MW: 148.1619 g/mol
MDL Number: MFCD11869778
CAS Number: 756839-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR