2-Methoxy-6-methylbenzonitrile ≥95%

Supplier: Matrix Scientific

074687-1G 074687-5G 074687-10G
102537-700EA 533.85 USD
102537-700 102537-702 102537-704
2-Methoxy-6-methylbenzonitrile ≥95%
2-Methoxy-6-methylbenzonitrile

Hazard: Irritant

Formula: C₉H₉NO
MW: 147.1739 g/mol
MDL Number: MFCD10699539
CAS Number: 53005-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR