2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)pyridine

074683-1G 074683-5G 074683-10G
102537-680EA 640 USD
102537-680 102537-682 102537-684
2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester ≥95%
2-(Tetrahydro-2H-pyran-4-yloxy)pyridine-5-boronic acid pinacol ester

Hazard: Irritant

Formula: C₁₆H₂₄BNO₄
MW: 305.18 g/mol
MDL Number: MFCD09702390
CAS Number: 910036-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR