4-Bromo-N-cyclohexylpyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

074682-250MG 074682-1G
102537-676EA 826.15 USD
102537-676 102537-678
4-Bromo-N-cyclohexylpyrimidin-2-amine ≥95%
4-Bromo-N-cyclohexylpyrimidin-2-amine

Hazard: Irritant

Formula: C₁₀H₁₄BrN₃
CAS Number: 1269292-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR