(4-Bromopyrimidin-2-yl)cyclopentylamine ≥95%

Supplier: Matrix Scientific

074681-250MG 074681-1G
102537-672EA 826.15 USD
102537-672 102537-674
(4-Bromopyrimidin-2-yl)cyclopentylamine ≥95%
(4-Bromopyrimidin-2-yl)cyclopentylamine

Hazard: Irritant

Formula: C₉H₁₂BrN₃
CAS Number: 1269291-43-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR