5-Methoxy-2-fluorobenzylcyanide ≥95%
Supplier: Matrix Scientific
074679-1G
074679-5G
102537-662EA
115.38
USD
102537-662
102537-664
5-Methoxy-2-fluorobenzylcyanide ≥95%
5-Methoxy-2-fluorobenzylcyanide
Hazard: Irritant
Formula:
C9H8FNO |
CAS Number:
672931-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...