Methyl-3-fluoro-4-methylbenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-fluoro-4-methylbenzoate

074678-1G 074678-5G
102537-656EA 218.46 USD
102537-656 102537-658
Methyl-3-fluoro-4-methylbenzoate ≥95%
Methyl 3-fluoro-4-methylbenzoate

Hazard: Irritant

Formula: C₉H₉FO₂
MW: 168.1650 g/mol
MDL Number: MFCD06203786
CAS Number: 87808-48-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR