(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride ≥95%

Supplier: Matrix Scientific

074677-1G 074677-5G
102537-652EA 710.77 USD
102537-652 102537-654
(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride ≥95%
(4-(4-Chlorophenoxy)phenyl)methanamine hydrochloride

Hazard: Irritant

Formula: C₁₃H₁₃Cl₂NO
MW: 270.1544 g/mol
MDL Number: MFCD02089410
CAS Number: 262862-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR